KEY465193427 Key Organics / BIONET

4-(4-formylpyrimidin-2-yl)phenyl 4-fluorobenzoate

  • MDL number MFCD02185964
  • Cas Number : 477870-91-0
  • Molecular Weight : 322.295

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65413

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0150_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart