KEY465193438 Key Organics / BIONET

4-{4-[(1E)-2-cyano-3-methoxy-3-oxoprop-1-en-1-yl]pyrimidin-2-yl}phenyl thiophene-2-carboxylate

  • MDL number MFCD02185971
  • Cas Number : 477871-08-2
  • Molecular Weight : 391.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65438

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0195_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart