KEY465193439 Key Organics / BIONET

(2E)-N-[(4-chlorophenyl)methyl]-3-[4-(3-fluoropropoxy)phenyl]prop-2-enamide

  • MDL number MFCD02102399
  • Cas Number : 1164506-13-1
  • Molecular Weight : 347.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65441

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0198_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart