KEY465193452 Key Organics / BIONET

(3E,5E)-1-benzyl-3,5-bis(phenylmethylidene)piperidin-4-ol

  • MDL number MFCD02185979
  • Cas Number : 477871-34-4
  • Molecular Weight : 367.492

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65473

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0614_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart