KEY465193456 Key Organics / BIONET

9-benzyl-1,6-diphenyl-9-azadispiro[2.1.2^{5}.3^{3}]decan-4-one

  • MDL number MFCD02570869
  • Cas Number : 477768-10-8
  • Molecular Weight : 393.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65479

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0622_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart