KEY465193459 Key Organics / BIONET

1-(4-methylbenzenesulfonyl)-2-nitro-4-(trifluoromethyl)benzene

  • MDL number MFCD00111089
  • Cas Number : 127291-63-8
  • Molecular Weight : 345.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65484

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0631_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart