KEY465193460 Key Organics / BIONET

methyl 4-(4-methylbenzenesulfonyl)-3-nitrobenzoate

  • MDL number MFCD02083003
  • Cas Number : 477871-40-2
  • Molecular Weight : 335.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65485

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0633_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart