KEY465193461 Key Organics / BIONET

ethyl 3,5-dimethyl-1-[5-(thiophene-2-amido)pyridin-2-yl]-1H-pyrazole-4-carboxylate

  • MDL number MFCD02570870
  • Cas Number : 477710-45-5
  • Molecular Weight : 370.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65487

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-0636_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart