KEY465193469 Key Organics / BIONET

(2Z)-2-[(2-fluorophenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-yl 4-nitrobenzoate

  • MDL number MFCD02083016
  • Cas Number : 477871-54-8
  • Molecular Weight : 382.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65506

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0656_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart