KEY465193470 Key Organics / BIONET

2-[(2-fluorophenyl)methyl]-1-azabicyclo[2.2.2]octan-3-one

  • MDL number MFCD02083019
  • Cas Number : 477871-59-3
  • Molecular Weight : 233.286

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65510

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0662_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart