KEY465193472 Key Organics / BIONET

4-(thiophen-2-yl)-1H,4H,5H,6H,7H-imidazo[4,5-c]pyridine

  • MDL number MFCD02571313
  • Cas Number : 64403-28-7
  • Molecular Weight : 205.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65515

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-0671_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart