KEY465193473 Key Organics / BIONET

2,4-dibromo-6-[(1E)-[(2-methoxyphenyl)imino]methyl]phenol

  • MDL number MFCD00125791
  • Cas Number : 329936-52-9
  • Molecular Weight : 385.055

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65517

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0674_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart