KEY465193478 Key Organics / BIONET

3'-(thiophen-2-yl)-3,5-dihydro-1H-spiro[2-benzothiepine-4,2'-oxirane]-5-one

  • MDL number MFCD02102420
  • Cas Number : 477871-63-9
  • Molecular Weight : 288.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65524

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0683_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart