KEY465193480 Key Organics / BIONET

1aH,2H,7H,7aH-naphtho[2,3-b]oxirene-2,7-dione

  • MDL number MFCD00044300
  • Cas Number : 15448-58-5
  • Molecular Weight : 174.155

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65529

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0689_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart