KEY465193483 Key Organics / BIONET

2,4-dibromo-6-{[(2-methoxyphenyl)amino]methyl}phenol

  • MDL number MFCD02102433
  • Cas Number : 477871-71-9
  • Molecular Weight : 387.071

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65541

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-0704_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart