KEY465193487 Key Organics / BIONET

3'-(2-fluorophenyl)-3-oxo-1-azaspiro[bicyclo[2.2.2]octane-2,2'-oxiran]-1-ium-1-olate

  • MDL number MFCD02102436
  • Cas Number : 477871-74-2
  • Molecular Weight : 263.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65547

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0711_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart