KEY465193489 Key Organics / BIONET

methyl 2-(1,1-dioxo-1lambda6-thiomorpholin-4-yl)-3-(1H-indol-3-yl)propanoate

  • MDL number MFCD02571315
  • Cas Number : 477871-76-4
  • Molecular Weight : 336.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65549

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0714_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart