KEY465193493 Key Organics / BIONET

methyl 2-{4-hydroxy-1-[(4-methylphenyl)methyl]-2-oxo-1,2-dihydropyridin-3-yl}acetate

  • MDL number MFCD02570879
  • Cas Number : 477871-82-2
  • Molecular Weight : 287.315

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65558

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-0808_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart