KEY465193494 Key Organics / BIONET

methyl 1-[2-(1H-indol-3-yl)ethyl]-2-methyl-5-oxo-4,5-dihydro-1H-pyrrole-3-carboxylate

  • MDL number MFCD02570880
  • Cas Number : 477871-83-3
  • Molecular Weight : 298.342

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65560

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0810_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart