KEY465193497 Key Organics / BIONET

2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-N-(morpholin-4-yl)acetamide

  • MDL number MFCD03011800
  • Cas Number : 383147-70-4
  • Molecular Weight : 323.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65563

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-0815_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart