KEY465193498 Key Organics / BIONET

4-[4-methyl-2-(pyridin-2-yl)-1,3-thiazol-5-yl]pyrimidin-2-amine

  • MDL number MFCD28347903
  • Cas Number : 499796-01-9
  • Molecular Weight : 269.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 255 - 258
Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/453049

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-0820_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart