KEY465193502 Key Organics / BIONET

ethyl 2-(4-methyl-3-oxo-3,4-dihydroquinoxalin-2-yl)acetate

  • MDL number MFCD02185980
  • Cas Number : 14152-57-9
  • Molecular Weight : 246.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65570

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-0828_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart