KEY465193509 Key Organics / BIONET

N-(3,5-dichlorophenyl)-2-(1-methyl-1H-pyrrol-2-yl)-2-oxoacetamide

  • MDL number MFCD02102450
  • Cas Number : 477871-97-9
  • Molecular Weight : 297.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65580

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-1009_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart