KEY465193512 Key Organics / BIONET

methyl 4-[2-(1-methyl-1H-pyrrol-2-yl)-2-oxoacetamido]benzoate

  • MDL number MFCD02102452
  • Cas Number : 477872-00-7
  • Molecular Weight : 286.287

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65583

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-1012_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart