KEY465193514 Key Organics / BIONET

1-(morpholin-4-yl)-2-(2-phenyl-1H-indol-3-yl)ethane-1,2-dione

  • MDL number MFCD02102454
  • Cas Number : 102003-06-5
  • Molecular Weight : 334.375

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65585

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-1016_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart