KEY465193521 Key Organics / BIONET

1-(2-phenyl-1H-indol-3-yl)-2-(piperidin-1-yl)ethane-1,2-dione

  • MDL number MFCD02102461
  • Cas Number : 102318-27-4
  • Molecular Weight : 332.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65592

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-1024_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart