KEY465193524 Key Organics / BIONET

4-methyl-N'-(4-methylbenzenesulfonyl)-1,2,3-thiadiazole-5-carbohydrazide

  • MDL number MFCD02102464
  • Cas Number : 477872-09-6
  • Molecular Weight : 312.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65595

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-1030_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart