KEY465193537 Key Organics / BIONET

3,4-dichloro-5-phenyl-2,5-dihydrofuran-2-one

  • MDL number MFCD00728660
  • Cas Number : 72857-85-3
  • Molecular Weight : 229.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Store at 4°C
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65611

Safety Information

signalword Warning
pcodes P411, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-1065_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart