KEY465193538 Key Organics / BIONET

1-(2,4-dichlorobenzenesulfonyl)-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD02102485
  • Cas Number : 477872-33-6
  • Molecular Weight : 327.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65624

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-1080_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart