KEY465193539 Key Organics / BIONET

tert-butyl 4-[(2,4-difluorophenyl)amino]piperidine-1-carboxylate

  • MDL number MFCD04124702
  • Cas Number : 860788-24-5
  • Molecular Weight : 312.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65642

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-1140_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart