KEY465193542 Key Organics / BIONET

3-{[(3-chlorophenyl)methyl]sulfanyl}-5-(thiophen-2-yl)-4H-1,2,4-triazol-4-amine

  • MDL number MFCD02186713
  • Cas Number : 477872-64-3
  • Molecular Weight : 322.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65671

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-1242_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart