KEY465193547 Key Organics / BIONET

N-[3-sulfanyl-5-(thiophen-2-yl)-4H-1,2,4-triazol-4-yl]thiophene-2-carboxamide

  • MDL number MFCD08064715
  • Cas Number : 923106-17-6
  • Molecular Weight : 308.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65677

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-1250_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart