KEY465193550 Key Organics / BIONET

5-methoxy-2-[3-(trifluoromethyl)phenyl]pyrimidin-4-ol

  • MDL number MFCD04124708
  • Cas Number : 860788-30-3
  • Molecular Weight : 270.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65711

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-1524_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart