KEY465193559 Key Organics / BIONET

(3E)-4-[6-(4-methylphenyl)imidazo[2,1-b][1,3]thiazol-5-yl]but-3-en-2-one

  • MDL number MFCD03787263
  • Cas Number : 860788-31-4
  • Molecular Weight : 282.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65753

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-0203_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart