KEY465193566 Key Organics / BIONET

2-{[5-(2-oxo-2-phenylethoxy)naphthalen-1-yl]oxy}-1-phenylethan-1-one

  • MDL number MFCD03011819
  • Cas Number : 319482-15-0
  • Molecular Weight : 396.442

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65773

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-0244_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart