KEY465193567 Key Organics / BIONET

4-(2-fluorophenyl)-2-methyl-5-oxo-1H,4H,5H-indeno[1,2-b]pyridine-3-carbonitrile

  • MDL number MFCD03008773
  • Cas Number : 371137-43-8
  • Molecular Weight : 316.335

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65774

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-0246_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart