KEY465193571 Key Organics / BIONET

2-[(furan-2-yl)methylidene]-2,3-dihydro-1H-indene-1,3-dione

  • MDL number MFCD00219001
  • Cas Number : 29874-41-7
  • Molecular Weight : 224.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65793

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-0281_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart