KEY465193575 Key Organics / BIONET

2-chloro-3-(pyrrolidin-1-yl)-1,4-dihydronaphthalene-1,4-dione

  • MDL number MFCD00010368
  • Cas Number : 59641-25-7
  • Molecular Weight : 261.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65800

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-0295_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart