KEY465193582 Key Organics / BIONET

3-(3-fluorophenyl)-3-(2,2,2-trifluoroacetamido)propanoic acid

  • MDL number MFCD03011834
  • Cas Number : 117291-14-2
  • Molecular Weight : 279.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65834

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-0610_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart