KEY465193584 Key Organics / BIONET

methyl 6-(4-bromophenyl)-2,4-dioxopiperidine-3-carboxylate

  • MDL number MFCD03102801
  • Cas Number : 860788-46-1
  • Molecular Weight : 326.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65836

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1T-0613_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart