KEY465193585 Key Organics / BIONET

5-{[1-(4-bromophenyl)-1H-pyrrol-2-yl]methylidene}-1,3-dimethyl-1,3-diazinane-2,4,6-trione

  • MDL number MFCD02789673
  • Cas Number : 428458-95-1
  • Molecular Weight : 388.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65838

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-0616_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart