KEY465193591 Key Organics / BIONET

methyl 3-(2-fluorophenyl)-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoate

  • MDL number MFCD03011840
  • Cas Number : 383148-35-4
  • Molecular Weight : 313.328

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65856

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1T-0639_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart