KEY465193593 Key Organics / BIONET

3,3-dimethyl-2,4-dioxa-9lambda6-thiaspiro[5.5]undecane-1,5,9,9-tetrone

  • MDL number MFCD03012087
  • Cas Number : 64096-86-2
  • Molecular Weight : 262.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65859

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-0642_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart