KEY465193595 Key Organics / BIONET

1-(5-chlorothiophen-2-yl)-3-(dimethylamino)but-2-en-1-one

  • MDL number MFCD03617982
  • Cas Number : 860788-50-7
  • Molecular Weight : 229.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65864

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1T-0650_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart