KEY465193597 Key Organics / BIONET

3-(3-bromophenyl)-3-(1,1-dioxo-1lambda6-thiomorpholin-4-yl)propanoic acid

  • MDL number MFCD03012205
  • Cas Number : 439095-27-9
  • Molecular Weight : 362.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65867

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-0653_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart