KEY465193603 Key Organics / BIONET

methyl (2E)-3-[(2,3-dihydro-1,4-benzodioxin-6-yl)amino]-2-(1H-1,2,4-triazol-1-yl)prop-2-enoate

  • MDL number MFCD03011848
  • Cas Number : 383148-51-4
  • Molecular Weight : 302.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65889

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-0826_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart