KEY465193604 Key Organics / BIONET

2-[(E)-[(2-nitrophenyl)methylidene]amino]-4,5-diphenylfuran-3-carbonitrile

  • MDL number MFCD03012532
  • Cas Number : 454199-52-1
  • Molecular Weight : 393.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65892

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1T-0830_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart