KEY465193605 Key Organics / BIONET

5'-[(E)-[(2E)-3-[4-(dimethylamino)phenyl]prop-2-en-1-ylidene]amino]-2'-(furan-2-yl)-[2,3'-bifuran]-4'-carbonitrile

  • MDL number MFCD03012533
  • Cas Number : 551930-71-3
  • Molecular Weight : 397.434

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65893

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-0831_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart