KEY465193606 Key Organics / BIONET

(3E)-4-(3,4,5-trimethoxyphenyl)but-3-en-2-one

  • MDL number MFCD01570023
  • Cas Number : 19039-94-2
  • Molecular Weight : 236.267

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65895

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-0833_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart