KEY465193609 Key Organics / BIONET

9,10-dimethoxy-1H,2H,3H,4H,6H,7H,11bH-pyrido[2,1-a]isoquinoline-2,4-dione

  • MDL number MFCD03305515
  • Cas Number : 5911-65-9
  • Molecular Weight : 275.304

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65908

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1T-0849_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart